missing translation for 'onlineSavingsMsg'
Learn More
Learn More
9-Bromo-1-nonene, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
It's employed as an important intermediate for raw material for organic synthesis, agrochemical, pharmaceutical and dyestuff field
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It′s employed as an important intermediate for raw material for organic synthesis, agrochemical, pharmaceutical and dyestuff field
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
It′s employed as an important intermediate for raw material for organic synthesis, agrochemical, pharmaceutical and dyestuff field
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications
Specifications
Color | Colorless to Yellow |
Boiling Point | 100°C to 101°C (15 mmHg) |
Linear Formula | Br(CH2)7CH=CH2 |
Refractive Index | 1.4675 |
Quantity | 1 g |
Beilstein | 1746338 |
Formula Weight | 205.14 |
Percent Purity | 97% |
Physical Form | Liquid |
Chemical Name or Material | 9-Bromo-1-nonene |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction