missing translation for 'onlineSavingsMsg'
Learn More

9-Bromo-10-(1-naphthyl)anthracene, 98%, Thermo Scientific™

Product Code. 15401049
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 400607-04-7
Molecular Formula C24H15Br
Molecular Weight (g/mol) 383.288
MDL Number MFCD11046571
InChI Key SYACRXBYRNYMLN-UHFFFAOYSA-N
Synonym 9-bromo-10-naphthalen-1-yl anthracene, 9-bromo-10-1-naphthalenyl anthracene, 9-bromo-10-1-naphthyl anthracene, anthracene, 9-bromo-10-1-naphthalenyl, 9-bromo-10-naphthalen-1-yl-anthracene, 10-bromo-9-phthalen-1-yl anthracene, 10-bromo-9-naphthalen-1-yl anthracene, 9-bromo-10-1-naphthalenyl-anthracene, 10-naphthalene-1-yl-9-bromo anthracene
PubChem CID 21076365
IUPAC Name 9-bromo-10-naphthalen-1-ylanthracene
SMILES C1=CC=C2C(=C1)C=CC=C2C3=C4C=CC=CC4=C(C5=CC=CC=C53)Br

Specifications

Color Yellow
Quantity 250 mg
Beilstein 20386637
Formula Weight 383.29
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 9-Bromo-10-(1-naphthyl)anthracene
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'