missing translation for 'onlineSavingsMsg'
Learn More

9-Bromo-10-phenylanthracene, 98%, Thermo Scientific Chemicals

Product Code. 15422159
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It is used as an OLED material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as an OLED material.

Solubility
Insoluble in water. Soluble in hot toluene, dimethylformamide.

Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23674-20-6
Molecular Formula C20H13Br
Molecular Weight (g/mol) 333.228
MDL Number MFCD00230983
InChI Key WHGGVVHVBFMGSG-UHFFFAOYSA-N
Synonym 10-bromo-9-phenylanthracene, anthracene, 9-bromo-10-phenyl, 9-bromo-10-phenyl-anthracene, ksc201k3d, 9-bromo-10-phenyl anthracene
PubChem CID 4155836
IUPAC Name 9-bromo-10-phenylanthracene
SMILES C1=CC=C(C=C1)C2=C3C=CC=CC3=C(C4=CC=CC=C42)Br

Specifications

Melting Point 154°C to 158°C
Color Yellow
Quantity 1 g
Beilstein 2277064
Solubility Information Insoluble in water. Soluble in hot toluene,dimethylformamide.
Formula Weight 333.23
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 9-Bromo-10-phenylanthracene

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.