missing translation for 'onlineSavingsMsg'
Learn More

9-Bromoanthracene, 96%, Thermo Scientific Chemicals

Product Code. 11386976
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

9-Bromoanthracene acts as an intermediate in the preparation of 9-substituted derivative of the polycyclic aromatic hydrocarbon (PAH) anthracene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
9-Bromoanthracene acts as an intermediate in the preparation of 9-substituted derivative of the polycyclic aromatic hydrocarbon (PAH) anthracene.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1564-64-3
Molecular Formula C14H9Br
Molecular Weight (g/mol) 257.13
MDL Number MFCD00001243
InChI Key ZIRVQSRSPDUEOJ-UHFFFAOYSA-N
Synonym anthracene, 9-bromo, 9-bromo anthracene, 9-bromo-anthracene, 9-anthracenyl bromide, 9-bromanthracen, pubchem9183, 9-bromoanthrene, acmc-209ddx, 9-bromoanthracene, dsstox_cid_29080
PubChem CID 74062
IUPAC Name 9-bromoanthracene
SMILES C1=CC=C2C(=C1)C=C3C=CC=CC3=C2Br

Specifications

Melting Point 97°C to 103°C
Quantity 5 g
Beilstein 1869747
Solubility Information Insoluble in water.
Formula Weight 257.14
Percent Purity 96%
Chemical Name or Material 9-Bromoanthracene

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.