missing translation for 'onlineSavingsMsg'
Learn More
Learn More
9-Bromophenanthrene, 96%, Thermo Scientific Chemicals
41.61 € - 177.89 €
Chemical Identifiers
CAS | 573-17-1 |
---|---|
Molecular Formula | C14H9Br |
Molecular Weight (g/mol) | 257.13 |
MDL Number | MFCD00001174 |
InChI Key | RSQXKVWKJVUZDG-UHFFFAOYSA-N |
Synonym | 9-phenanthryl bromide, phenanthrene, 9-bromo, 9-phenathryl bromide, 9-bromo-phenanthrene, 9-bromo phenanthrene, 9-bromophenathrene, 9-bromanylphenanthrene, pubchem13993, acmc-1b0zz, 9-bromophenanthrene |
PubChem CID | 11309 |
IUPAC Name | 9-bromophenanthrene |
SMILES | C1=CC=C2C(=C1)C=C(C3=CC=CC=C23)Br |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10390494
|
Thermo Scientific Acros
107240100 |
10 g | Glass bottle |
41.61 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10745941
|
Thermo Scientific Acros
107240250 |
25 g | Glass bottle |
62.31 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10776321
|
Thermo Scientific Acros
107241000 |
100 g | Glass bottle |
177.89 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
573-17-1 | |
257.13 | |
RSQXKVWKJVUZDG-UHFFFAOYSA-N | |
11309 | |
C1=CC=C2C(=C1)C=C(C3=CC=CC=C23)Br |
C14H9Br | |
MFCD00001174 | |
9-phenanthryl bromide, phenanthrene, 9-bromo, 9-phenathryl bromide, 9-bromo-phenanthrene, 9-bromo phenanthrene, 9-bromophenathrene, 9-bromanylphenanthrene, pubchem13993, acmc-1b0zz, 9-bromophenanthrene | |
9-bromophenanthrene |
Specifications
573-17-1 | |
100.0 | |
Yellow | |
>110°C | |
95% min. (GC) | |
MFCD00001174 | |
9-phenanthryl bromide, phenanthrene, 9-bromo, 9-phenathryl bromide, 9-bromo-phenanthrene, 9-bromo phenanthrene, 9-bromophenathrene, 9-bromanylphenanthrene, pubchem13993, acmc-1b0zz, 9-bromophenanthrene | |
C1=CC=C2C(=C1)C=C(C3=CC=CC=C23)Br | |
257.13 | |
257.13 | |
Powder |
95.0 | |
60°C to 65°C | |
180°C to 190°C (2.0 mmHg) | |
Authentic | |
C14H9Br | |
05, 671 | |
RSQXKVWKJVUZDG-UHFFFAOYSA-N | |
9-bromophenanthrene | |
11309 | |
96% | |
9-Bromophenanthrene, 96% |
Safety and Handling
missing translation for 'einecsNumber' : 209-351-6
missing translation for 'rtecsNumber' : SF7197000
Product Identifier
- 9-Bromophenanthrene
RUO – Research Use Only