missing translation for 'onlineSavingsMsg'
Learn More

9-Fluorenone, 99+%

Product Code. 11904851 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass Bottle
Plastic bottle
Unit Size:
100g
2.5kg
500g
5g
This item is not returnable. View return policy

Product Code. 11904851

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 486-25-9 | C13H8O | 180.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 486-25-9
InChI Key YLQWCDOCJODRMT-UHFFFAOYSA-N
Synonym 9-fluorenone, 9h-fluoren-9-one, fluorenone, 9-oxofluorene, diphenylene ketone, unii-az9t83s2aq, ccris 593, 9h-fluorene-9-one, az9t83s2aq, 9-fluoreneone
PubChem CID 10241
ChEBI CHEBI:17922
IUPAC Name fluoren-9-one
SMILES C1=CC=C2C(=C1)C3=CC=CC=C3C2=O

Specifications

Melting Point 81.0°C to 85.0°C
Color Yellow
Boiling Point 342.0°C
Flash Point 163°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Plastic bottle
Molecular Formula C13H8O
Quantity 2.5 kg
Beilstein 07,465
Solubility Information Solubility in water: insoluble.
Molecular Weight (g/mol) 180.21
Formula Weight 180.21
Percent Purity 99+%
Physical Form Crystalline Powder or Flakes
Chemical Name or Material 9-Fluorenone
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.