missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 9-Hydroxy-9-fluorenecarboxylic acid, 97%

Product Code. 11490527
Click to view available options
Quantity:
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 467-69-6
Molecular Formula C14H10O3
Molecular Weight (g/mol) 226.231
MDL Number MFCD00001137
InChI Key GXAMYUGOODKVRM-UHFFFAOYSA-N
Synonym flurenol, 9-hydroxy-9-fluorenecarboxylic acid, flurecol, 9-hydroxy-9h-fluorene-9-carboxylic acid, glycolic acid, diphenylene, 9h-fluorene-9-carboxylic acid, 9-hydroxy, flurecol bsi:iso, unii-4tkt65am8p, 9-hydooxyfluorene-9-carboxylic acid, 9-hydroxy-9-fluorene carboxyli acid
PubChem CID 10087
ChEBI CHEBI:82015
IUPAC Name 9-hydroxyfluorene-9-carboxylic acid
SMILES C1=CC=C2C(=C1)C3=CC=CC=C3C2(C(=O)O)O

Specifications

Melting Point ∼164°C (decomposition)
Quantity 5 g
UN Number UN3077
Beilstein 1314711
Formula Weight 226.23
Percent Purity 97%
Chemical Name or Material 9-Hydroxy-9-fluorenecarboxylic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'