missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals 9-Hydroxyrisperidone, 98% Product Code.: 15335068

Thermo Scientific Chemicals 9-Hydroxyrisperidone, 98%

Product Code. 15335068
1 g
Click to view available options
Quantity:
1 g
This item is not returnable. View return policy

Product Code. 15335068

Brand: Thermo Scientific Chemicals 456250010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 144598-75-4
Melting Point 171.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Molecular Formula C23H27FN4O3
Quantity 1 g
Synonym paliperidone, 9-hydroxyrisperidone, invega, 9-oh-risperidone, invega sustenna, xeplion, palliperidone, 3-2-4-6-fluoro-1,2-benzisoxazol-3-yl-1-piperidinyl ethyl-6,7,8,9-tetrahydro-9-hydroxy-2-methyl-4h-pyrido 1,2-a pyrimidin-4-one, chembl1621, 3-2-4-6-fluorobenzo d isoxazol-3-yl piperidin-1-yl ethyl-9-hydroxy-2-methyl-6,7,8,9-tetrahydro-4h-pyrido 1,2-a pyrimidin-4-one
InChI Key PMXMIIMHBWHSKN-UHFFFAOYSA-N
SMILES CC1=C(C(=O)N2CCCC(C2=N1)O)CCN3CCC(CC3)C4=NOC5=C4C=CC(=C5)F
IUPAC Name 3-[2-[4-(6-fluoro-1,2-benzoxazol-3-yl)piperidin-1-yl]ethyl]-9-hydroxy-2-methyl-6,7,8,9-tetrahydropyrido[1,2-a]pyrimidin-4-one
Molecular Weight (g/mol) 426.48
PubChem CID 115237
ChEBI CHEBI:83804
Formula Weight 426.48
Percent Purity 98%
Chemical Name or Material 9-Hydroxyrisperidone
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0350378 Certificate TypeCertificate of Analysis Date04/03/2022 Catalog Number456250010
compliance-icons
Product Identifier
  • 9-Hydroxyrisperidone
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals 9-Hydroxyrisperidone, 98% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.