missing translation for 'onlineSavingsMsg'
Learn More
9-Methylanthracene, 99%, Thermo Scientific Chemicals Product Code.: 10011160

9-Methylanthracene, 99%, Thermo Scientific Chemicals

Product Code. 10011160
5 g, Glass bottle
Click to view available options
Quantity:
5 g
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10011160

Brand: Thermo Scientific Acros 126300050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 779-02-2
Molecular Formula C15H12
Molecular Weight (g/mol) 192.26
MDL Number MFCD00001261
InChI Key CPGPAVAKSZHMBP-UHFFFAOYSA-N
Synonym anthracene, 9-methyl, unii-65nk4cin03, ccris 2740, 9-methyl anthracene, 9-methyl-anthracene, anthracen-9-ylmethyl, acmc-209pcm, 9-methylanthracene, wln: l c666j b1, 9-methylanthracene, analytical standard
PubChem CID 13068
IUPAC Name 9-methylanthracene
SMILES CC1=C2C=CC=CC2=CC2=CC=CC=C12

Specifications

Melting Point 76.0°C to 79.0°C
Color Orange to Yellow
Density 1.0660g/mL
Boiling Point 196.0°C to 197.0°C (12.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Quantity 5 g
Beilstein 05,II,586
Specific Gravity 1.066
Formula Weight 192.26
Percent Purity 99%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 9-Methylanthracene
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberR05J029 Certificate TypeCertificate of Analysis Date05/09/2023 Catalog Number126300500,126300100,126300050
Lot NumberA0441428 Certificate TypeCertificate of Analysis Date26/04/2022 Catalog Number126300500,126300100,126300050
Lot NumberA0426162 Certificate TypeCertificate of Analysis Date03/02/2021 Catalog Number126300500,126300100,126300050
Lot NumberA0393869 Certificate TypeCertificate of Analysis Date18/08/2020 Catalog Number126300500,126300100,126300050
Lot NumberA0418917 Certificate TypeCertificate of Analysis Date05/05/2020 Catalog Number126300500,126300100,126300050
compliance-icons compliance-icons
Product Identifier
  • 9-Methylanthracene
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
9-Methylanthracene, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.