missing translation for 'onlineSavingsMsg'
Learn More

9-(Trimethylsilyl)fluorene, 99%, Thermo Scientific Chemicals

Product Code. 11444847
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7385-10-6
Molecular Formula C16H18Si
Molecular Weight (g/mol) 238.41
MDL Number MFCD00092570
InChI Key YFPRGLHBYSNHMW-UHFFFAOYSA-N
Synonym 9-trimethylsilyl fluorene, 9h-fluoren-9-yl trimethyl silane, 9-trimethylsilylfluorene, silane, 9h-fluoren-9-yltrimethyl, 9-fluorenyl trimethylsilane, 9h-fluoren-9-yl trimethyl silane #
PubChem CID 553570
IUPAC Name 9H-fluoren-9-yl(trimethyl)silane
SMILES C[Si](C)(C)C1C2=CC=CC=C2C3=CC=CC=C13

Specifications

Melting Point 95°C to 98°C
Quantity 5 g
Formula Weight 238.4
Percent Purity 99%
Chemical Name or Material 9-(Trimethylsilyl)fluorene

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'