Learn More
Uvex™ 9169 Super F OTG CR Safety Spectacles
21.65 EUR valid until 2024-07-31
Use promo code "21674" to get your promotional price.
Use promo code "21674" to get your promotional price.
Click to view available options
Clear
Description
- Designed to provide professinal protection in laboratories, clinics, cleanrooms, or in food production
- Spectacles are capable of withstanding multiple autoclave sterilization cycles
- Can be worn over perscription lenses, making them perfect for visitors
- Certified to EN 166 (personal eye protection) and EN 170 (UV filters)
- Uvex™ Supravision Clean lens coating features a chemical-resistant, anti-fog interior and an extremely scratch-resistant exterior
- Soft arm ends provide additional comfort
- Unrestricted peripheral vision
Specifications
Lens Type | Supravision Clean |
Certifications/Compliance (Europe) | EN 166:2001, EN 170:2002 |
Lens Tint | Clear |
Frame Color | White, Blue |
Lens Material | Polycarbonate |
UV Protection | UV400 |
Autoclavable | Autoclavable |
Labeling | W 166 FT CE — 2C-1.2 W 1 FKN |