Learn More
uvex 9169 Super F Over-the-Glasses Safety Glasses
These extremely comfortable safety spectacles provide excellent eye area protection, and fit perfectly over conventional prescription lenses. Specs are available in a variety of lens coating and lens tints.
19.85 € - 22.50 €
Specifications
Model 9169 Super F OTG
Material Polycarbonate
Disposable Reusable
Lens Material Polycarbonate
Frame Material Plastic
View More Specs
Products 3
Product Code Brand Lens Coating Lens Tint Price Quantity & Availability  
Product Code Brand Lens Coating Lens Tint Price Quantity & Availability  
17217261
View Documents
uvex
Anti-fog, Scratch-resistant Clear
19.85 €
Each
Estimated Shipment: 17-01-2025
to see stock.
Add to basket
17207261
View Documents
uvex
Anti-fog, Scratch-resistant Light Brown
22.50 €
Each
Estimated Shipment: 31-01-2025
to see stock.
Add to basket
17237261
View Documents
uvex
Scratch-resistant Amber This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
  • Overspecs for users who also wear prescription lenses with an arm-hinge concept
  • Side arms graduate to soft, flexible material for enhanced comfort and grip
  • Unrestricted peripheral vision
Supravision Excellence Coating
  • Inside: Anti-fog
  • Outside: Scratch-resistant and chemical-resistant
  • Easy to clean, and less susceptible to soiling
Supravision Sapphire Coating
  • Inside: Scratch-resistant and chemical-resistant
  • Outside: Scratch-resistant and chemical-resistant
  • Non-stick properties allow for easy removal of greasy/watery dirt
Infradur Plus Coating
  • Inside: Anti-fog
  • Outside: Scratch-resistant
  • Minimizes burns due to welding sparks
Specifications
Specifications
9169 Super F OTG
Reusable
Plastic
Polycarbonate
Polycarbonate
Standard