missing translation for 'onlineSavingsMsg'
Learn More
Learn More
uvex 9190 i-3 Replacement Lens
Click to view available options
Clear
Description
- Clear, i-3 replacement lens
- Supravision excellence lens coating
Specifications
Specifications
Lens Type | Supravision excellence |
Lens Coating | Anti-fog |
Certifications/Compliance (Europe) | EN 166:2001, EN 170:2002 |
Lens Tint | Clear |
Product Type | Ergonomic Safety Glasses |
Model | 9190 i-3 |
Material | Polycarbonate |
Disposable | Reusable |
Lens Material | Polycarbonate |
Frame Material | Plastic |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction