missing translation for 'onlineSavingsMsg'
Learn More

uvex Pheos Safety Spectacles

A soft component connected directly to lens provides optimum protection against dust and water. The innovative X-tended Eyeshield adapts to the wearer's face for enhanced comfort, with X-Twist side arm technology for a secure fit.

12.40 € - 30.50 €

Specifications

Product Type Safety Glasses
Material Polycarbonate
Disposable Reusable
Lens Material Polycarbonate
Frame Material Plastic
View More Specs

Products 9
Product Code Brand Lens Coating Lens Tint Frame Color Price Quantity & Availability  
Product Code Brand Lens Coating Lens Tint Frame Color Price Quantity & Availability  
17267301
View Documents
uvex
Anti-fog Silver Mirror Blue, Gray
30.50 €
Each
Estimated Shipment: 31-01-2025
to see stock.
Add to basket
 
17257301
View Documents
uvex
Anti-fog, Scratch-resistant Amber Black, Yellow
16.65 €
Each
Estimated Shipment: 31-01-2025
to see stock.
Add to basket
 
17217301
View Documents
uvex
Anti-fog, Scratch-resistant Clear Black, Light Blue
13.95 €
Each
Estimated Shipment: 31-01-2025
to see stock.
Add to basket
 
17227301
View Documents
uvex
Anti-fog, Scratch-resistant Clear Blue, Gray
13.95 €
Each
Estimated Shipment: 31-01-2025
to see stock.
Add to basket
 
17237301
View Documents
uvex
Anti-fog, Scratch-resistant Clear Red, Gray
16.15 €
Each
Estimated Shipment: 31-01-2025
to see stock.
Add to basket
 
17207301
View Documents
uvex
Anti-fog, Scratch-resistant Gray White, Black
15.35 €
Each
Estimated Shipment: 31-01-2025
to see stock.
Add to basket
 
17297291
View Documents
uvex
Anti-fog, Scratch-resistant Light Brown White, Black
16.65 €
Each
Estimated Shipment: 17-01-2025
to see stock.
Add to basket
 
17247301
View Documents
uvex
Scratch-resistant Clear Blue, Gray
12.40 €
Each
Estimated Shipment: 31-01-2025
to see stock.
Add to basket
 
15366082
View Documents
uvex
Anti-fog, Scratch-resistant Clear White, Lime Green
12.90 €
Each
Estimated Shipment: 21-01-2025
to see stock.
Add to basket
 
Description

Description

  • Safety spectacles with a modern, sporty design
  • Duo-spherical lens provides a large field of vision
  • Can easily be combined with other forms of PPE

Supravision Excellence Coating

  • Interior: Anti-fog
  • Exterior: Scratch-resistant and chemical-resistant
  • Lenses are easy to clean and less susceptible to soiling

Supravision Sapphire Coating

  • Inside: Scratch-resistant and chemical-resistant
  • Outside: Scratch-resistant and chemical-resistant
  • Non-stick properties allow for easy removal of greasy/watery dirt
Specifications

Specifications

Safety Glasses
Reusable
Plastic
Standard
Polycarbonate
Polycarbonate
UV400
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.