missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ 96-Well, Cell Culture-Treated, Flat-Bottom, Half-Area Microplate
Shop All Corning Products

Click to view available options
Packaging:
20/Pk., 100/Cs.
4/Pk
Description
Greener Choice Claims
- Environmental benefits include:
- Certifications and Seals of Approval
- Energy Star Certification
- Renewable Energy

Specifications
Specifications
Format | Half-Area |
Lid | With Lid |
Array | 8 x 12 |
Packaging | 4/Pk |
Binding Type | TC-Treated |
Color | Black, Clear Bottom |
For Use With (Equipment) | For high content cell-based assays using imaging systems |
Material | Cycloolefin Copolymer |
No. of Wells | 96 |
Sterility | Sterile |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Corning™ 96-Well, Cell Culture-Treated, Flat-Bottom, Half-Area Microplate >
Spot an opportunity for improvement?Share a Content Correction