missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One 96-well, Cell Culture-Treated, Flat-Bottom Microplate

Product Code. 16670364
Click to view available options
Packaging:
1/Pk., 16/Cs.
This item is not returnable. View return policy
This item is not returnable. View return policy

Color:Black, Clear Bottom; Sterility:Sterile; Lid:With Lid

  • Features black pigmented frame with 190μm ultraclear film bottom for ideal compatibility with instrument optics
  • Universal microscope objective compatibility
  • Refractive index similar to glass (1.53)
  • Chimney well design minimizes risk of cross contamination

Service Plan

No

Warranty

No

TRUSTED_SUSTAINABILITY

Specifications

Format Standard
Lid With Lid
Array 8 x 12
Packaging 1/Pk., 16/Cs.
Color Black, Clear Bottom
Material Cycloolefin
No. of Wells 96
Sterility Sterile
Surface Treatment Cell Culture
Well Shape Flat Bottom (F-well)
No. per Case 16
Quantity 16/Cs.
Well Color Clear
Well Design Chimney
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'