Learn More
Fisherbrand™ 96-Well Low-Profile, Skirted PCR Plates, Natural
Fit most thermal cyclers
Brand:  Fisherbrand™ 14230241
 View more versions of this product
Product Code. 12194112
  • 113.00 € / Pack of 25
Estimated Shipment: 07-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Fit most thermal cyclers
  • Virgin polypropylene
  • Clear well bottom for sample visibility
  • Stackable
  • Raised-rim design
  • Sealable using adhesive films and foils, heat seals, and cap strips
  • 200μL maximum well volume when used with adhesive and heat seals
  • Cut-away corner and alphanumeric grid
  • SBS (Society for Biomolecular Screening) footprint
  • Automation friendly
  • Eight holes in the skirt aid plate positioning and removal from thermal cycler block
  • Cleanroom produced
Specifications
Specifications
Standard
Yellow
Skirted
200 μL
PCR, RT-PCR applications
Clear
8 x 12
PCR Microplate
Polypropylene
96
Yes