missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ 96-Well Microtiter™ Microplates
Use these 96-well microplates for micro-volume applications such as storage, sample or reagent transfer, serial dilution, hemagglutination, blood typing and compliment fixation.
99.35 € - 261.77 €
Specifications
Format | Standard |
---|---|
Array | 8 x 12 |
Color | Clear |
Product Type | Microplate |
No. of Wells | 96 |
Product Code | Brand | Material | Well Shape | Volume (Metric) Well | Lid | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Material | Well Shape | Volume (Metric) Well | Lid | Price | Quantity & Availability | ||
10558295
|
Thermo Scientific™
2205 |
Polystyrene | Round Bottom (U-well) | 400 μL | Without Lid |
99.35 €
Pack of 50 |
In Stock
Log in to see stock. |
||
10437305
|
Thermo Scientific™
2605 |
Polystyrene | Conical Bottom (V-well) | 280 μL | Without Lid |
108.00 €
Pack of 50 |
In Stock
Log in to see stock. |
||
10519465
|
Thermo Scientific™
9205 |
Polystyrene | C | 330 μL | Without Lid |
137.00 €
Pack of 50 |
In Stock
Log in to see stock. |
||
10222275
|
Thermo Scientific™
2101 |
Vinyl | Round Bottom (U-well) | 400 μL | Without Lid |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
10567145
|
Thermo Scientific™
2801 |
Vinyl | Flat Bottom (F-well) | 330 μL | Without Lid |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
10324165
|
Thermo Scientific™
2401 |
Vinyl | Round Bottom (U-well) | 400 μL | With Lid |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
Description
Thermo Scientific™ 96-Well Microtiter Microplates are ideal for micro-volume applications such as storage, sample or reagent transfer, serial dilution, hemagglutination, blood typing and compliment fixation.
- For high-throughput screening and research
- Clear plates for colorimetric applications
- Standard footprint, 128 × 86mm
- Available as both polystyrene square wells and vinyl square wells
- Vinyl Microtiter microplates are used for complement fixation, agglutination, and blood typing
- Styrene Microtiter microplates are used for complement fixation, agglutination, microbiology testing, and blood typing
Ordering Information:
Polystyrene microplates are available as 50 units/package. Vinyl microplates are available as 25 or 100 units/package, depending on product.
Specifications
Standard | |
Clear | |
96 |
8 x 12 | |
Microplate | |
Non-treated |