missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BTX™ HT 200 Plate Handler, Auto-Sense Track Switching
Designed to work with 96 well plates. BTX™ Harvard Apparatus 96 Well Plate Handler helps to create a safe and reliable working area for processing electroporation plates.
Brand: BTX™ 450401
This item is not returnable.
View return policy
Description
- Quickly optimize for better gene delivery
- Designed to work with 96 well plates
- Pulse switching technology integrated into package and gold plated contacts to mate with electroporation plate
- With an adapter to accommodate 25 well plates as well with our ECM 630 generator
- Prosess up to 96 wells in seconds
Specifications
25-well, 96-well | |
ECM630, ECM830, Gemini X2 | |
Electroporation accessory | |
HT-200 Microplate Handler, Auto-Sense Track Switching | |
Pin | |
1 to 99 | |
5°C to 40°C | |
20 to 80% | |
4.8 kg | |
Banana Cables | |
Exponential Decay Wave, Square Wave |
High-Throughput Electroporation | |
BTX Electroporation | |
3000 VDC, 250 VAC | |
LCD | |
Auto sense track switching, Connectors for standard pulse generator | |
10 μs to 10 s | |
1 Ea. | |
2 mm, 4 mm | |
Non-sterile | |
Digital User | |
Yes |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction