missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eppendorf™ 96-Well Protein LoBind Deep Well Plates
Use these plates in proteomics or other fields of protein research, where protein concentration tend to be very small and sample recovery is vital for assay results. Eppendorf™ Deepwell Protein LoBind Plates recover significantly more protein for downstream analyses and enzymes remain active.
Brand: Eppendorf™ 0030504100
740.39 EUR valid until 2024-12-31
Use promo code "21693" to get your promotional price.
This item is not returnable.
View return policy
Description
- LoBind material guarantees maximum sample recovery for improved assay results
- Free of surface coating (e.g., silicone) to minimize the risk of sample interference
- Lot-certified PCR clean purity grade: of human DNA, DNAase, RNase and PCR inhibitors
- Available in tube, microplate, and deepwell plate formats for easy-up scaling
- Unique OptiTrack™ matrix: 30% faster sample identification and fewer pipetting errors
- RecoverMax™ well design: optimized well geometry for minimal remaining/dead volume and excellent mixing properties
- Raised well rims and a smooth surface guarantee reliable sealing in plates
Specifications
Includes Barcode | |
Standard | |
Autoclavable | |
White | |
Polypropylene | |
96 | |
Round Bottom (U-well) | |
Sample storage and preparation, bacteria and yeast cultivation, DNA and RNA isolation with high throughput | |
Clear |
SBS, IVD, EN ISO 9001/2/3 and EN 13485 guidelines | |
8 x 12 | |
40 Plates (5 Bags x 8 Plates) | |
Protein LoBind Plate | |
500 μL | |
Non-sterile | |
Yes | |
40 Pk. |