missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Abgene™ 96-well Sealing Mats for Sample Processing & Storage DeepWell™ and MicroWell Microplates
GREENER_CHOICE

Round or square CapMats, Sealing Mats, MatCaps for sealing 96-well storage plates, available in autoclavable and piercable formats.

Brand:  Thermo Scientific™ AB-0566/SP

 View more versions of this product

Product Code. 10722684

  • 182.00 € / Pack of 50
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Round or square CapMats, Sealing Mats, MatCaps for sealing 96-well storage plates, available in autoclavable and piercable formats.

Thermo Scientific 96-well Sealing Mats are designed for 96-well storage plates and are available in either piercable or autoclavable formats. The round or square capped mats are made of thermoplastic elastomer and are suitable for long-term storage.

Highlights

  • Certified RNAse, DNAse, PCR Inhibitor, and human DNA free
  • Round or square capped mats for sealing 96-well storage plates
  • Alphanumeric grid-referencing
  • Molded from a proprietary thermoplastic elastomer for a flexible design
  • Suitable for long term storage
  • Available in autoclavable or piercable formats

Pierceable version:

  • Solvent resistant (including DMSO, Ethanol, and Isopropanol)
  • Suitable for Gamma Irradiation
  • Requires 6N for piercing
  • Temperature Range: −20°C to +55°C

Autoclavable version:

  • Suitable for Gamma Irradiation
  • Temperature Range: −20°C to +130°C

 

Hazardous: No
Storage Condition: Ambient

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Natural
CapMat for 96-well Polypropylene Sample Processing & Storage Plates
TPE
Autoclavable
50
7mm
Designed for sealing 96-well storage plate and suitable for long term storage
96-well capmat, sealing mat
None
Yes
Round
PCR Ready
Alcohol Resistant, Solvent Resistant
Certified RNase, DNase, PCR Inhibitors, and human DNA Free. ANSI Compliant
Cleanroom
96
10
50 Mats
96-well
Abgene
Yes
-20°C to +120°C
Abgene 96 Round Well Plate
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers