missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ twin.tec™ PCR Plates LoBind™ - PCR Plates
GREENER_CHOICE

Product Code. 15911992 Shop All Eppendorf Products
Click to view available options
Color:
Blue
Clear
Green
Orange
Red
Yellow
No. of Wells:
96
384
Volume (Metric) Well:
45 μL
150 μL
250 μL
Packaging:
10 Bags x 30 Plates
300 Plates
5 Bags x 5 Plates
Unit Size:
Pack of 25
Pack of 300
This item is not returnable. View return policy

Product Code. 15911992

Brand: Eppendorf™ 0030129547

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Eppendorf twin.tec™ PCR plates LoBind, available in 96- and 384-well formats, maximize nucleic acid recovery and are certified as PCR clean for consistently reliable results.

  • LoBind material guarantees maximum sample recovery for improved assay results.
  • One-piece design: combining a polycarbonate frame and polypropylene wells for optimum performance.
  • Extremely thin-walled polypropylene wells guarantee optimum heat transfer to the sample.
  • Exceptionally solid and torque-resistant polycarbonate frame.
  • OptiTrack™ matrix for faster sample identification and fewer pipetting errors.
  • Raised well rims for effective sealing, also reduces risk of cross-contamination.
  • Batch-tested and certified free of DNA, DNAase, RNase and PCR inhibitors (PCR clean).
TRUSTED_SUSTAINABILITY

Tekniske data

Format Standard
Array 16 x 24
Autoclavable Autoclavable
Packaging 5 Bags x 5 Plates
Color Clear
Product Type PCR Plate
Skirt Style Skirted
Material Polypropylene, Polycarbonate
Volume (Metric) Well 45 μL
No. of Wells 384
Sterility Non-sterile
Surface Treatment LoBind
Dnase Free Yes
Max. RCF 6000 x g
No. per Pack 25
Purity or Quality Grade PCR Clean (Non-sterile)
Quantity 25 Pk.
RNase Free Yes
Well Color Colorless
Vis mere Vis mindre
Korrektion af produktindhold

Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.

Produkttitel

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.

Mange tak! Din feedback er blevet sendt. Hos Fisher Scientific arbejder vi altid på at forbedre vores indhold for dig. Vi sætter pris på din feedback.