missing translation for 'onlineSavingsMsg'
Learn More

Uvex™ 9906 Pheos SLB1 Polycarbonate Visors

Visors are treated with supravision excellence coating for anti-fog and anti-scratch protection. Certified in accordance with GS-ET 29 as Class 1 Visors for protection from electrical arcs (4 kA).

111.00 € - 130.00 €

Specifications

Certifications/Compliance EN 166:2001, EN 170:2002
Color Black
For Use With (Equipment) Pheos E-S-WR Safety Helmets
Lens Tint Clear
Material Polycarbonate
View More Specs

Products 2
Product Code Brand Mounting Type Price Quantity & Availability  
Product Code Brand Mounting Type Price Quantity & Availability  
17280272
View Documents
Uvex™
9906006
Magnetic Locks
130.00 €
Each
Estimated Shipment: 30-12-2024
to see stock.
Add to basket
 
17270272
View Documents
Uvex™
9906005
Mechanical Levers
111.00 €
Each
Estimated Shipment: 30-12-2024
to see stock.
Add to basket
 
Description

Description

  • Magnetic locking system is extremely easy to attach to the visor
  • Ergonomic joint system is comfortable to wear, even for long periods
  • Certified in accordance with GS-ET 29 as Class 1 Visors for protection from electrical arcs (4 kA)
  • Labeling: W 166 3 8-2 9 B CE 0196 (Frame), 2C-1.2 W 1 B 8-1-0 9 KN CE 0196 (Lens)
  • Compatible only with Pheos E-S-WR helmets
  • Lens can be changed
  • Helmet sold separately

Supravision Excellence Coating

  • Interior: Anti-fog
  • Exterior: Scratch-resistant and chemical-resistant
  • Lenses are easy to clean and less susceptible to soiling
Specifications

Specifications

EN 166:2001, EN 170:2002
Pheos E-S-WR Safety Helmets
Polycarbonate
Visor
Black
Clear
9906 Pheos SLB1
Class 1
Videos
SDS
Documents

Documents

Product Certifications