missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ 9mm Non-Assembled Plastic Screw Thread Wide Opening Autosampler Vial Kits

Save time and reduce the risk of contamination using this convenience kit. Each kit includes 100 plastic vials and 100 screw thread caps with pre-assembled septa.

56.80 € - 65.70 €

Specifications

No. per Pack 100
Product Line Surestop
Closure Size 9 mm
Marking Spot No
View More Specs

Products 2
Product Code Brand Closure Size Material Septum Price Quantity & Availability  
Product Code Brand Closure Size Material Septum Price Quantity & Availability  
15206583
View Documents
Thermo Scientific™
C5000-87
9 mm Polypropylene, PTFE, Silicone Red PTFE/White Silicone This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15276583
View Documents
Thermo Scientific™
C5000-97
9 mm Polypropylene, PTFE, Silicone Blue PTFE/White Silicone This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Save time during sample preparation and reduce the risk of contamination using Thermo Scientific™ 9mm Wide Opening Plastic Vial Convenience Kits (Unassembled). Each kit includes 100 plastic vials and 100 screw thread caps with pre-assembled septa. Reusable two-compartment trays protect vials and closures while keeping matching supplies together. Clear trays to make it easy to keep track of available supplies without opening containers. Choose from Thermo Scientific™, Thermo Scientific™ National™, and Thermo Scientific™ Chromacol™ brands.

Save time during sample preparation and reduce the risk of contamination using Thermo Scientific™ 9mm Wide Opening Plastic Vial Convenience Kits (Unassembled). Each kit includes 100 plastic vials and 100 screw thread caps with pre-assembled septa. Reusable two-compartment trays protect vials and closures while keeping matching supplies together. Clear trays to make it easy to keep track of available supplies without opening containers. Choose from Thermo Scientific™, Thermo Scientific™ National™, and Thermo Scientific™ Chromacol™ brands.
Specifications

Specifications

100
Surestop
9 mm
No
Videos
SDS
Documents

Documents

Product Certifications

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.