missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SI Analytics™ A 1180 pH Glass Electrode
SI Analytics™ A 1180 pH Glass Electrode ensures high measurement accuracy and stability and long service life.
Brand: SI Analytics™ 1057997
This item is not returnable.
View return policy
Description
- Ag / AgCl discharge
- A Glass
- Very deep vessels
- Proven high-end electrodes for demanding measurement tasks
- Utmost versatility of pH electrodes is achieved by a large selection of junctions, membrane glass types and shapes, shaft lengths and diameters, ground joints, plug connections and integrated temperature sensors
- Comes with individual serial number and quality certificate
- Silamid™ reference system
Specifications
Glass Stem | |
pH Electrode | |
Spherical | |
120 mm |
0 to 14 | |
Plug Head | |
0°C to 80°C | |
12 mm |