missing translation for 'onlineSavingsMsg'
Learn More
A 839977, Tocris Bioscience™ Product Code.: 18095092

A 839977, Tocris Bioscience™

Product Code. 18095092
10 mg
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy

Product Code. 18095092

Brand: Tocris Bioscience 4232/10

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Potent P2X7 antagonist

Potent P2X7 antagonist; blocks BzATP-evoked calcium influx at recombinant human, rat and mouse P2X7 receptors (IC50 values are 20, 42 and 150 nM respectively). Displays antinociceptive effects in rat and mice models of inflammatory pain. CNS penetrant.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 870061-27-1
Molecular Formula C19H14Cl2N6O
Molecular Weight (g/mol) 413.262
InChI Key GMVNBKZQJFRFAR-UHFFFAOYSA-N
Synonym 1-2,3-dichlorophenyl-n-2-pyridin-2-yloxy phenyl methyl-1,2,3,4-tetrazol-5-amine, d0m0xc, 1-2,3-dichlorophenyl-n-2-pyridin-2-yloxy benzyl-1h-tetrazol-5-amine, 1-2,3-dichlorophenyl-n-2-2-pyridinyloxy phenyl methyl-1h-tetrazol-5-amine, 1-2,3-dichlorophenyl-n-2-pyridin-2-yloxy phenyl methyl-1h-1,2,3,4-tetrazol-5-amine
PubChem CID 53325875
IUPAC Name 1-(2,3-dichlorophenyl)-N-[(2-pyridin-2-yloxyphenyl)methyl]tetrazol-5-amine
SMILES C1=CC=C(C(=C1)CNC2=NN=NN2C3=C(C(=CC=C3)Cl)Cl)OC4=CC=CC=N4

Specifications

Quantity 10 mg
Formula Weight 413.26
Percent Purity >98%
Chemical Name or Material A 839977
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
A 839977, Tocris Bioscience™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.