Learn More
Thermo Scientific™ ARCTIC A10 Refrigerated Circulators
A10 refrigerated bath circulators provide efficient cooling for open and closed applications, with temperatures ranging from -10° to +100°C and 6L maximum bath volume.
Brand: Thermo Scientific™ 1535101
3746.61 EUR valid until 2024-12-31
Use promo code "21916" to get your promotional price.
Description
Thermo Scientific™ A10 refrigerated bath circulators provide efficient cooling for open and closed applications, with temperatures ranging from -10° to +100°C and maximum bath volume of 6 liters. Powerful bath circulators with a stainless-steel reservoir can be configured with the controller that best meets circulation and temperature ranges for your application.
Optional force/suction pumps and sophisticated digital control allow you to ensure accuracy and reproducibility. Choose from the STANDARD, ADVANCED or PREMIUM controllers to create the perfect fit for your application.- Energy-saving feature limits power consumption: tight control of cooling capacity dependent on application needs
- Small refrigeration circulators are optimized for small footprint and low noise
- Robust stainless-steel baths
- The controller can be turned to 90° on all sides of the bath for optimal viewing
- External circulation set included for plumbing connection M16×1, 0.25 NPT male, 0.375in. O.D.
- Three-side ventilation concept: up to two sides can be blocked with full refrigeration performance and no degradation of equipment
- Draining port at the front
- Integrated handles
- CFC-free refrigerant
Includes:
Communication cables, bridge with gasket and thumbscrews, fittings, clamps, pump plug for external circulation, 6ft. power cord, bath cover
Recommended for:
- Calibration
- Bioreactors
- Rotary Evaporators
- Condensers
- Sample/Material Testing
- Sample/Material Preparation
Specifications
1.58 gal. | |
6 L | |
RS232, RS485, Ethernet/LAN, Analog I/O | |
17 L/min. | |
-10°C to +100°C | |
3/FL | |
16.3 x 8.7 x 24.9 in. (41.4 x 22 x 63.2 cm) | |
4.9 in. | |
12.4 cm | |
5.4 in. | |
13.7 cm | |
63.2 cm | |
15 cm | |
27.5 kg | |
230 V |
SC150 | |
CE/ROHS/WEEE | |
240 W | |
Force Pump, Suction Pump | |
300 mbar/4.35 psi | |
0.02°C | |
16.3 in. | |
41.4 cm | |
8.7 in. | |
22 cm | |
24.9 in. | |
5.9 in. | |
60.6 lb. | |
230 V 50 Hz | |
50 Hz |