missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ A12B4C3
Potent and selective polynucleotide kinase/phosphatase (PNKP) inhibitor
351.00 € - 1380.00 €
Specifications
Purity | >98% |
---|---|
Product Type | A12B4C3 |
Molecular Weight (g/mol) | 534.65 |
Inhibitors | A12B4C3 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18710118
|
Tocris Bioscience™
5097/10 |
10 mg |
351.00 €
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18728788
|
Tocris Bioscience™
5097/50 |
50 mg |
1380.00 €
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Potent and selective polynucleotide kinase/phosphatase (PNKP) inhibitor (IC50 = 60 nM). Displays no effect on calcineurin, protein PP-1 or APTX. Interrupts DNA repair and enhances radiosensitivity of human A549 lung carcinoma and MDA-MB-231 cells.Specifications
>98% | |
A12B4C3 | |
534.65 | |
A12B4C3 |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title