missing translation for 'onlineSavingsMsg'
Learn More

Kimberly-Clark Professional™ KleenGuard™ A40 Liquid and Particle Protection Coveralls

Product Code. 11774903
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Improves safety and comfort in high-risk areas providing general protection against water-based or chemical splashes. Kimberly-Clark™ KleenGuard™ A40 Liquid and Particle Protection Coveralls have the added benefit of anti-static protection.

  • White in color
  • Single use, disposable
  • Silicone-free and anti-static
  • Specially designed hood to fit a respirator
  • Helps to reduce contamination in low lint and critical areas
  • Full-length zip, over-locked stitched seams and double-layer fabric
  • Ideal for protection against larger particles, radioactive dust and chemical splashes
  • Suited for pharmaceutical areas, manufacturing, utilities, electronics, agriculture and paint-spraying areas

Specifications

Product Type General Purpose Coveralls
Size 3X-Large
Gender Unisex
Material KleenGuard™
Color White
Disposable Single-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Hood Type Attached
Closure Type Front Zipper
Seam Type Stitched
Sterility Sterile
Resistance Water, Chemicals, Liquid, Particle, Anti-static
Latex Free Yes
Length (English) 15.6 in.
Length (Metric) 40 cm
Weight (English) 18.282 lb.
Weight (Metric) 8.31 kg
Quantity 25/Pk.
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'