Learn More
Honeywell™ A69 Peak Series Hard Hat
Multipurpose helmet with ribbed ABS outer shell and 6-point nylon web suspension.
12.38 € - 23.20 €
Specifications
Color White
Includes Adjustable headband and suspension
Material Shell: ABS, Suspension: Nylon web
Style Ribbed, rain trough
View More Specs
Products 2
Product Code Brand Suspension Type Price Quantity & Availability  
Product Code Brand Suspension Type Price Quantity & Availability  
15886795
View Documents
Honeywell™
933170
6-point nylon web with pin-lock adjustment This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15896795
View Documents
Honeywell™
933180
6-point nylon web with ratchet adjustment
23.20 €
Each
Estimated Shipment: 06-01-2025
to see stock.
Add to basket
Description
Description
  • Rain trough design
  • Selection of pin-lock and ratchet suspensions
  • Size and headband height adjustable to ensure wearer comfort
  • Slots to fit face shield and earmuffs
  • Easy sweatband replacement system
  • Personalizing with logo printing available
  • ABS shell
Specifications
Specifications
White
Adjustable headband and suspension
Shell: ABS, Suspension: Nylon web
Ribbed, rain trough