missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ AA 147
ER proteostasis regulator
120.00 € - 443.00 €
Specifications
Chemical Name or Material | N-(2-Hydroxy-5-methylphenyl)benzenepropanamide |
---|---|
CAS | 393121-74-9 |
Molecular Formula | C16H17NO2 |
Formula Weight | 255.31 |
Purity | 98% |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18737386
|
Tocris Bioscience™
6759/25 |
25 mg |
443.00 €
25mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18075912
|
Tocris Bioscience™
6759/5 |
5 mg |
120.00 €
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Endoplasmic reticulum (ER) proteostasis regulator
- Activates the ATF6 transcriptional program independently of ER stress and/or global UPR activation
- Selectively reduces secretion of amyloidogenic transthyretin (TTR) from HepG2 cells
- Reduces amyloidogenic immunoglobulin light chain (ALLC) secretion from ALMC-2 cells (EC50 = 1.1 μM).
Specifications
N-(2-Hydroxy-5-methylphenyl)benzenepropanamide | |
C16H17NO2 | |
98% |
393121-74-9 | |
255.31 |
Safety and Handling
Recommended Storage : Store at 4°C
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title