missing translation for 'onlineSavingsMsg'
Learn More

Atago™ Abbe NAR-1T Liquid Refractometer

Designed for liquid sample measurement. Atago™ Abbe NAR-1T Liquid Refractometers feature the Refractive Index Scale and the BRIX scale.

Brand:  Atago™ 1211

Product Code. 10719200

  • 3555.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Includes a D line (589nm) light source
  • Calibration can be done with distilled water
Specifications

Specifications

Refractometer
±0.0002nD
For Liquid Sample Measurement
130 x 180 x 230 mm
130 mm
5°C to 50°C
240 V
1.3 to 1.7
Digital
1 Each
230 mm
180 mm
240 V 50/60 Hz
50/60 Hz
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.