Learn More
ATAGO™ Abbe Refractometer NAR-2T
The Abbe Refractometer NAR-2T is designed for use with compounds which require measurement at a high temperature.
Brand:  ATAGO™ 1220
Product Code. 11486238
  • 5330.00 € / Each
Estimated Shipment: 21-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Abbe table refractometer for the measurement of refractive index (1.3000 to 1.7000 ND) and Brix (0.0% to 95.0% scale) of high temperature samples between 5°C to 120°C and measures with thermometer unit from 0.0°C to 120°C.
  • LED illumination digital display of temperature readings in Celcius, and immediate analog scale display measurement readings with a D (deuterium) line (589 Nm) fiber optic light source
  • Operates on AC 100V to 240V, 50/60 Hz power supply, has a 110V power connection cord with a 6 ft. power cord, and can be calibrated with distilled water for ease of operation
  • Vertical, angled body (12 x 20 x 25 cm) with monocular, focusable, padded eyepiece for viewing
  • Can be connected to a constant temperature circulating bath (sold separately) for sample temperature control
Specifications
Specifications
Benchtop Refractometer
1.3000 to 1.7000
Measuring Compounds at High Temperatures
120 x 200 x 250 mm (Refractometer), 100 x 110 x 70 mm (Thermometer)
240 V
0.0% to 95.0% (Brix)
±0.0002 (Refractive Index), ±0.1% (Brix)
2.8 kg (Refractometer), 0.5 kg (Thermometer)
5°C to 120°C
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products