missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ AbC™ Anti-Rat/Hamster Bead Kit

AbC™ Anti-Rat/Hamster Bead Kit

Brand:  Molecular Probes™ A10389

243.21 EUR valid until 2025-06-30
Use promo code "24074" to get your promotional price.


Product Code. 10389194

  • 363.00 € / 1 set

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description
Show More
Specifications

Specifications

Contains 1 vial AbC™ anti-rat/hamster capture beads and 1 vial of negative beads.
  • Store at 2°C to 8°C
  • 6 μm
    Flow Cytometer
    1 kit
    Surface
    Compensation Beads
    Fluorescence
    any lasers (UV to 633/635)
    Tube
    Room Temperature
    CompenFlow™, Molecular Probes™
    Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents

    Documents

    One moment while we fetch your results.
    Certificates
    Special Offers

    Special Offers

    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title
    Molecular Probes™ AbC™ Anti-Rat/Hamster Bead Kit > 1 kit

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

    For Research Use Only. Not for use in diagnostic procedures.