missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AbC™ Total Antibody Compensation Bead Kit
Provides a consistent, accurate, and simple-to-use technique for the setting of flow cytometry compensation when using fluorochrome-conjugated mouse, rat, hamster, or rabbit antibodies with any laser in flow cytometry.
139.00 € - 415.00 €
Specifications
Content And Storage | Contains 1 vial AbC™ Total Compensation capture beads and 1 vial of negative beads. |
---|---|
Detection Method | Fluorescence |
Diameter (Metric) | 6 μm |
Product Line | AbC™ |
Product Type | Compensation Beads |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15308666
|
Invitrogen™
A10513 |
25 tests |
139.00 €
25 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15384147
|
Invitrogen™
A10497 |
100 tests |
415.00 €
100 tests |
In Stock
Log in to see stock. |
|||||
Description
- Broadest multispecies reactivity—for use with all isotypes of mouse, rat, hamster, and rabbit antibody conjugates
- Brightest signal—most sensitive antibody-capture beads available
- Maximum laser compatibility—for use with all lasers
- Each kit provides polystyrene microspheres that have fluorescent antibody conjugate capture capacity (positive compensation beads) or are inert (negative compensation beads).
- Ideal for compensation with all isotypes of mouse (IgG1, IgG2a, IgG2b, IgG3), rat (IgG1, IgG2a, IgG2b), hamster (Armenian IgG and Syrian IgG), and rabbit (monoclonal and polyclonal) immunoglobulins.Demonstrates the highest and most consistent reactivity to the different subclasses of immunoglobulin in the industry.
Specifications
Contains 1 vial AbC™ Total Compensation capture beads and 1 vial of negative beads. |
|
6 μm | |
Compensation Beads | |
Tube | |
Room Temperature |
Fluorescence | |
AbC™ | |
any lasers (UV to 633/635) | |
Flow Cytometer |
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title