Learn More
Thermo Scientific™ Abgene™ 96 Well 2.2mL Polypropylene DeepWell™ Sample Processing & Storage Plate
2.2 mL deepwell storage plates for the culture or storage of large samples.
Brand: Thermo Scientific™ AB-0932
314.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Thermo Scientific™ Abgene™ polypropylene microplates and deepwell plates offer storage security for assays, compound libraries or storing samples for either intermediate or long term use. Abgene microtiter plates are manufactured to exacting specifications in our Class 100,000 clean room ISO 9001 conditions using high quality medical grade virgin polypropylene resins, which ensure confidence in the quality and performance of the storage plates. Designed to ANSI standards these microplates and deepwell plates are compatible with a variety of automated liquid handling for high throughput workflows. The polypropylene storage plates are offered with spindle, pyramidal, U and V bottom wells along with a wide range of volumes to accommodate most applications.
Highlights- Designed for optimal sample recovery
- Each square well is specifically designed with an independent sealing rim to prevent cross-contamination
- U-bottom (AB-0661) to aid resuspension or conical bottom (AB-0932) for ease of sample retrieval
Specifications
Custom Barcoding Available | |
96-well | |
8 x 12 | |
Individually Wrapped | |
Abgene | |
96-well plate | |
Polypropylene | |
96 | |
Cleanroom | |
Square, Conical Bottom (V-well) | |
Automation Liquid Handlers, Biobanking, Genomics, Sample Prep, NGS, Sequencing, Long and Short Term Storage, Drug Discovery, Vaccine Research, PCR and qPCR sample preparation | |
50 | |
50/Cs. | |
Yes | |
ANSI/ SLAS Standard |
Certified RNase, DNase, PCR Inhibitors, and human DNA Free. ANSI Compliant | |
96-well Polypropylene Sample Processing & Storage Plates | |
Autoclavable | |
Low binding PP | |
Natural | |
ALPS5000, ALPS3000, and ALPS50V Heat Sealers, Automated Liquid Handlers, Multidrop Combi, Centrifuges, Ultralow Freezers, Liquid Handlers | |
2200 μL | |
1800 μL | |
Non-treated | |
Yes | |
5000 x g | |
1 | |
For Research Use Only. Not for use in diagnostic procedures. | |
PCR Ready |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.