Learn More
Thermo Scientific™ Abgene™ 96 Well 0.8mL Polypropylene DeepWell™ Sample Processing & Storage Plate for Genomics and NGS library preparation

0.8 mL DeepWell™ sample processing and storage plates for automation, NGS and genomic library prep, cell culture and sample storage applications. Specifically designed for use with liquid handlers, and magnetic bead extraction
Brand: Thermo Scientific™ AB-0859
181.44 EUR valid until 2025-05-31
Use promo code "25075" to get your promotional price.
Description
Thermo Scientific™ Abgene™ polypropylene microplates and DeepWell plates offer storage security for assays, compound libraries or storing samples for either intermediate or long term use and have molded slots and pads for automated sample processing and preparation workflows. Abgene microtiter plates are manufactured to exacting specifications in our Class 100,000 clean room ISO 9001 conditions using USP <661.1> high quality medical grade virgin polypropylene resins, which ensure confidence in the quality and performance of the storage plates. Designed to ANSI standards these microplates and DeepWell plates are compatible with a variety of automated liquid handling for high throughput workflows. The polypropylene storage plates are offered with spindle, pyramidal, U and V bottom wells along with a wide range of volumes to accommodate most applications.
All Thermo Scientific plates are manufactured in our cleanroom facility to ensure molecular grade quality. Our wide range of polypropylene plates provides excellent solutions for sample storage and assay set-up, allowing dilutions and aliquots to be handled, stored, or transported easily. The Virgin medical grade resin passes requirements for USP <661.1> and USP <665> reports have been completed for both CapMat sealed and Heat Sealed plates.
Highlights
- Certified free from DNAse, RNAse, PCR Inhibitor, and human DNA
- 0.8 mL maximum well volume when sealed with adhesive or heat seal
- 0.70 mL working volume when sealed with Storage Plate Cap Strip
- 0.55 mL working volume when sealed with a 96-well Sealing Mat
- Conical well bottom for ease of sample retrieval
Compatible Sealing: Adhesive and Heat seals
Hazardous: No
Max Centrifugal Force: 2000 g
Max Vol (Cap Seal): 0.7 mL
Max Vol (Film Seal): 0.8 mL
Max Vol (Mat Seal): 0.55 mL
Storage Condition: Ambient
All Thermo Scientific Abgene plates are manufactured in our cleanroom facility to ensure molecular grade quality. Our wide range of polypropylene plates provides excellent solutions for sample processing and storage and assay set-up, allowing dilutions and aliquots to be handled, stored, or transported easily.
All plates are ANSI format for compatibility with automated systems.
To further assist in sample storage and tracking, all plates can be customized with non-repeating barcoding and labelling. OEM options for custom logos, colors and full packaging support are available.
Certifications
Certified free from human DNA, RNases, and DNases.

Specifications
Custom Barcoding Available | |
Standard | |
8 x 12 | |
Bulk | |
Abgene | |
96-well plate | |
Polypropylene | |
96 | |
Cleanroom | |
Round, Conical Bottom (V-well) | |
Automation Liquid Handlers, Biobanking, Genomics, Sample Prep, NGS, Sequencing, Long and Short Term Storage, Drug Discovery, Vaccine Research, PCR and qPCR sample preparation | |
50 | |
For Research Use Only. Not for use in diagnostic procedures. | |
PCR Ready |
Certified RNase, DNase, PCR Inhibitors, and human DNA Free. ANSI Compliant | |
96-well Polypropylene Sample Processing & Storage Plates | |
Autoclavable | |
Low binding PP | |
Natural | |
ALPS5000, ALPS3000, and ALPS50V Heat Sealers, Automated Liquid Handlers, Multidrop Combi, Centrifuges, Ultralow Freezers, Liquid Handlers | |
800 μL | |
700 μL | |
Non-treated | |
Yes | |
2000 x g | |
50 | |
Yes | |
ANSI/ SLAS Standard |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
1424071 | Certificate of Compliance | 18/04/2025 | AB-0859 |
1423647 | Certificate of Compliance | 25/03/2025 | AB-0859 |
1422775 | Certificate of Compliance | 01/03/2025 | AB-0859 |
1422454 | Certificate of Compliance | 01/02/2025 | AB-0859 |
1420775 | Certificate of Compliance | 22/01/2025 | AB-0859 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.