missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Abgene™ 96 Well 0.8mL Polypropylene DeepWell™ Sample Processing & Storage Plate for Genomics and NGS library preparation
GREENER_CHOICE

0.8 mL DeepWell™ sample processing and storage plates for automation, NGS and genomic library prep, cell culture and sample storage applications. Specifically designed for use with liquid handlers, and magnetic bead extraction

Brand:  Thermo Scientific™ AB-0765/R

181.99 EUR valid until 2025-02-21
Use promo code "24115" to get your promotional price.


 View more versions of this product

Product Code. 10583215

  • 274.72 € / Pack of 50
Estimated Shipment: 01-04-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Thermo Scientific™ Abgene™ polypropylene microplates and DeepWell plates offer storage security for assays, compound libraries or storing samples for either intermediate or long term use and have molded slots and pads for automated sample processing and preparation workflows. Abgene microtiter plates are manufactured to exacting specifications in our Class 100,000 clean room ISO 9001 conditions using USP <661.1> high quality medical grade virgin polypropylene resins, which ensure confidence in the quality and performance of the storage plates. Designed to ANSI standards these microplates and DeepWell plates are compatible with a variety of automated liquid handling for high throughput workflows. The polypropylene storage plates are offered with spindle, pyramidal, U and V bottom wells along with a wide range of volumes to accommodate most applications.

All Thermo Scientific plates are manufactured in our cleanroom facility to ensure molecular grade quality. Our wide range of polypropylene plates provides excellent solutions for sample storage and assay set-up, allowing dilutions and aliquots to be handled, stored, or transported easily. The Virgin medical grade resin passes requirements for USP <661.1> and USP <665> reports have been completed for both CapMat sealed and Heat Sealed plates.

 

Highlights

  • Certified free from DNAse, RNAse, PCR Inhibitor, and human DNA
  • 0.8 mL maximum well volume when sealed with adhesive or heat seal
  • 0.70 mL working volume when sealed with Storage Plate Cap Strip
  • 0.55 mL working volume when sealed with a 96-well Sealing Mat
  • Conical well bottom for ease of sample retrieval

Compatible Sealing: Adhesive and Heat seals
Hazardous: No
Max Centrifugal Force: 2000 g
Max Vol (Cap Seal): 0.7 mL
Max Vol (Film Seal): 0.8 mL
Max Vol (Mat Seal): 0.55 mL
Storage Condition: Ambient

All Thermo Scientific Abgene plates are manufactured in our cleanroom facility to ensure molecular grade quality. Our wide range of polypropylene plates provides excellent solutions for sample processing and storage and assay set-up, allowing dilutions and aliquots to be handled, stored, or transported easily.

All plates are ANSI format for compatibility with automated systems.

To further assist in sample storage and tracking, all plates can be customized with non-repeating barcoding and labelling. OEM options for custom logos, colors and full packaging support are available.

Certifications

Certified free from human DNA, RNases, and DNases.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Custom Barcoding Available
96-well
8 x 12
Individually Wrapped
Abgene
96-well plate
Polypropylene
96
Non-treated
Yes
50
For Research Use Only. Not for use in diagnostic procedures.
PCR Ready
Certified RNase, DNase, PCR Inhibitors, and human DNA Free. ANSI Compliant
96-well Polypropylene Sample Processing & Storage Plates
Autoclavable
Low binding PP
Red
ALPS5000, ALPS3000, and ALPS50V Heat Sealers, Automated Liquid Handlers, Multidrop Combi, Centrifuges, Ultralow Freezers, Liquid Handlers
800 μL
Cleanroom
Round, Conical Bottom (V-well)
Automation Liquid Handlers, Biobanking, Genomics, Sample Prep, NGS, Sequencing, Long and Short Term Storage, Drug Discovery, Vaccine Research, PCR and qPCR sample preparation
1
Yes
ANSI/ SLAS Standard
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.