Learn More
Thermo Scientific™ Abgene™ 96 Well 1.2mL Polypropylene DeepWell™ Sample Processing & Storage Plate
Optimize suspension cell culture with these U-bottomed 1.2 mL storage plates.
Brand: Thermo Scientific™ AB-0564/K
Description
Thermo Scientific™ Abgene™ polypropylene microplates and deepwell plates offer storage security for assays, compound libraries or storing samples for either intermediate or long term use. Abgene microtiter plates are manufactured to exacting specifications in our Class 100,000 clean room ISO 9001 conditions using high quality medical grade virgin polypropylene resins, which ensure confidence in the quality and performance of the storage plates. Designed to ANSI standards these microplates and deepwell plates are compatible with a variety of automated liquid handling for high throughput workflows. The polypropylene storage plates are offered with spindle, pyramidal, U and V bottom wells along with a wide range of volumes to accommodate most applications.
ABgene Storage and Assay Plates are manufactured in a Class 100,000 cleanroom environment to ensure that each plate is contamination-free. Plates are made using virgin medical grade polypropylene and have highly polished surfaces, making them low binding and chemically resistant. Rigid side walls and industry standard footprint make plates suitable for automation and robotic handling, as well as for low-throughput applications.Plates feature square well geometry which promotes efficient sample aeration, making it ideal for bacterial growth applications. The low profile design maximizes centrifuge capacity, allowing more plates to be processed per run.
- 96 Square wells with 1.2mL maximum well volume
- Max. working volume: Film sealing = 1.2mL, Mat Cap Sealing = 1.0mL
- Square well format to maximize storage capacity
- Raised rim design around each well for optimal sealing and reduced risk of cross-contamination
- Suitable for use with DMSO, Ethanol, Isopropanol
- Alphanumeric grid-referencing
- Designed according to SBS footprint standards for robotic compatibility
- Wide working temperature range from -196° to +121°C (-321° to +250°F)
- Can be bar coded for improved sample tracking, please inquire
- Max. centrifugation speed: 2000g
- Autoclavable (15 min. at 121°C), safe for gamma irradiation sterilization
- Manufactured from virgin medical grade polypropylene and certified RNase-, DNase- and endotoxin-free
Compatible with:
Sealing options include: Adhesive Sealing (1.2mL working volume); Heat sealing (1.2mL working volume); 96 Cap Sealing Mats (1.0mL working volume)
Specifications
Custom Barcoding Available | |
96-well | |
8 x 12 | |
Individually Wrapped | |
Abgene | |
96-well plate | |
Polypropylene | |
96 | |
Non-treated | |
Yes | |
50 | |
For Research Use Only. Not for use in diagnostic procedures. | |
PCR Ready |
Certified RNase, DNase, PCR Inhibitors, and human DNA Free. ANSI Compliant | |
96-well Polypropylene Sample Processing & Storage Plates | |
Not Autoclavable | |
Low binding PP | |
Black | |
ALPS5000, ALPS3000, and ALPS50V Heat Sealers, Automated Liquid Handlers, Multidrop Combi, Centrifuges, Ultralow Freezers, Liquid Handlers | |
1200 μL | |
Cleanroom | |
Round | |
Automation Liquid Handlers, Biobanking, Genomics, Sample Prep, NGS, Sequencing, Long and Short Term Storage, Drug Discovery, Vaccine Research, PCR and qPCR sample preparation | |
1 | |
Yes | |
ANSI/ SLAS Standard |