Learn More
Thermo Scientific™ Abgene™ 96 Well Polypropylene Sample Processing & Storage Microplates

Streamline your molecular biology applications with these 96-well Polypropylene Storage Plates.
212.00 € - 398.00 €
Specifications
Barcode | Custom Barcoding Available |
---|---|
Certifications/Compliance | Certified RNase, DNase, PCR Inhibitors, and human DNA Free. ANSI Compliant |
Format | Standard |
Description | 96-well Polypropylene Sample Processing & Storage Plates |
Array | 8 x 12 |
Product Code | Brand | Well Shape | Volume (Metric) Well | Fit for Purpose | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Well Shape | Volume (Metric) Well | Fit for Purpose | Price | Quantity & Availability | ||||
11554754
![]() |
Thermo Scientific™
AB-0796 |
Round | 330 μL | PCR Ready |
212.00 €
Pack of 100 |
In Stock
Log in to see stock. ![]() |
||||
10304513
![]() |
Thermo Scientific™
AB-1058 |
Round, Conical Bottom (V-well) | 200 μL | PCR Ready |
398.00 €
Pack of 100 |
Estimated Shipment: 17-06-2025
Log in to see stock. |
||||
Description
Thermo Scientific Abgene 96 Well Polypropylene Storage Microplates provide excellent solutions for sample storage and assay set-up, allowing dilutions and aliquots to be handled, stored, or transported easily. All plates are ANSI format for compatibility with automated systems and are manufactured in our cleanroom facility to ensure molecular grade quality. To further assist in storage and tracking, all plates can be supplied with custom barcoding.
Features- Alphanumeric grid-referencing and industry standard footprint
- Polypropylene
- Sealing options: Adhesive Seals, Heat Seals and Cap Strips
- Cleanroom manufactured
Applications
- Sample storage
- Sample resuspension
- Cell culture
- Assay setup: diluting and aliquoting
Specifications
Custom Barcoding Available | |
Standard | |
8 x 12 | |
Bulk | |
Abgene | |
96-well plate | |
Polypropylene | |
Cleanroom | |
Yes | |
5000 x g | |
1 | |
Yes | |
ANSI/ SLAS Standard |
Certified RNase, DNase, PCR Inhibitors, and human DNA Free. ANSI Compliant | |
96-well Polypropylene Sample Processing & Storage Plates | |
Autoclavable | |
Low binding PP | |
Natural | |
ALPS5000, ALPS3000, and ALPS50V Heat Sealers, Automated Liquid Handlers, Multidrop Combi, Centrifuges, Ultralow Freezers, Liquid Handlers | |
96 | |
Non-treated | |
Automation Liquid Handlers, Biobanking, Genomics, Sample Prep, NGS, Sequencing, Long and Short Term Storage, Drug Discovery, Vaccine Research, PCR and qPCR sample preparation | |
100 | |
For Research Use Only. Not for use in diagnostic procedures. | |
PCR Ready |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.