Learn More
Testo™ 635-2 Temperature and Moisture Meter
Capable of measuring moisture and temperature of different types of materials, surfaces, and environments. Testo™ 635-2 Temperature and Moisture Meter works with a wide range of optional sensors.
Brand:  Testo™ 0563 6352
Product Code. 10047792
  • 994.00 € / Each
Explore more special offers
Description
Description
  • Optional sensors for moisture / temperature / pressure dew point / absolute pressure measurements
  • 2 plug-in and 3 wireless sensors
  • Super-size memory for up to 10,000 readings
  • USB cable
  • PC software for logging and archiving measuring data
Specifications
Specifications
0 to 100%
NTC: ±0.2°C (−25 - 74.9°C), K TC: ±0.3°C (−60 - 60°C)
ABS, TPE, Metal
220 x 74 x 46 mm
0.1%rH
-40°C, -150°C (NTC), -200°C to +1370°C (K TC)
0.1°C
428 g
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products