missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Ac-Cys-NHMe

Building block for the lysine acylation using conjugating enzymes (LACE) technique

153.00 € - 584.00 €

Specifications

Chemical Name or Material (2R)-2-(Acetylamino)-3-mercapto-N-methylpropanamide
CAS 10061-65-1
Target Unnatural Amino Acids
Molecular Formula C6H12N2O2S
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30000012
View Documents
Tocris Bioscience™
7936/10
10 mg
153.00 €
10mg
Please to purchase this item. Need a web account? Register with us today!
30000025
View Documents
Tocris Bioscience™
7936/50
50 mg
584.00 €
50mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Ac-Cys-NHMe is a non-canonical amino acid that is an essential building block for lysine acylation using conjugating enzymes (LACE) technique. LACE is a chemoenzymatic approach that uses an E2 small ubiquitin-like modifier (SUMO)-conjugating enzyme, Ubc9, to conjugate peptide thioesters containing a C-terminal ubiquitin-derived sequence (LRLRGG) to a lysine residue located in a recognition sequence (IKQE) on a protein via an isopeptide bond. Ac-Cys-NHMe can be reacted with a peptide hydrazide to form a reactive peptide thioester, which is the substrate for Ubc9. LACE enables the site-specific modification of proteins at lysine residues to allow attachment of biochemical probes such as fluorophores to proteins or to engineer proteins for site-specific delivery.
Specifications

Specifications

(2R)-2-(Acetylamino)-3-mercapto-N-methylpropanamide
10061-65-1
Unnatural Amino Acids
C6H12N2O2S
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.