Learn More
ACADM Mouse, anti-Human, CoraLite 488, Clone: 1A11G6, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-67742-100UL
This item is not returnable.
View return policy
Description
ACADM, also named as MCAD, belongs to the acyl-CoA dehydrogenase family. This enzyme is specific for acyl chain lengths of 4 to 16. It catalyzes the reaction: Acyl-CoA + acceptor = 2,3-dehydroacyl-CoA + reduced acceptor. Defects in ACADM are the cause of medium-chain acyl-CoA dehydrogenase deficiency (MCAD deficiency). This protein can exsit as a dimer(PMID:8962055). This antibody is specific to ACADM.
Specifications
ACADM | |
Monoclonal | |
CoraLite 488 | |
100 μL | |
34 | |
Liquid |
Immunofluorescence | |
1A11G6 | |
Mouse | |
Primary | |
Human |