Learn More
Thermo Scientific™ Access Key Options for Ultra-Low Temperature Freezers
Record who opened the ultra low freezer door, when it was opened and how long it was open with access key options.
Brand:  Thermo Scientific™ ACU34567
 View more versions of this product
Product Code. 11336381
  • 58.09 € / Pack of 5
Estimated Shipment: 21-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Analyze records of which staff opened the freezer door, what time is was opened and for what duration with the Thermo Scientific™ Access Key Options for Ultra-Low Temperature Freezers.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Access Key Pack (U.S.): Includes five cards, supports ISO15693 Protocol
Freezer Access Key
TSX Series, Forma 89000, HERAfreeze HLE, Revco RLE and TLE Series