Learn More
BINDER™ Right Access Port (10 mm)
For Use With: BINDER units KB 23, KB 53, KB 115
Brand:  BINDER™ 8012-0392
Product Code. 15681726
  • 386.00 € / Each
Estimated Shipment: 29-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • For use on right
  • Includes silicone plug
  • For use with a wide range of BINDER™ incubators and chambers
Specifications
Specifications
Access port with silicone plug - 10 mm, right
Silicone
Silicone plug
BINDER Chambers KB 23, KB 53, KB 115
Access Port
10 mm