Learn More
SymLine™ System Accessories
Dovetail your SymLine™ System to suit your specific needs! Here, you will find a lot of useful ancillaries.
Brand: SymLine™ 106609
This item is not returnable.
View return policy
Description
- SymLine™ offers many useful accessories such as manifolds, shut-off valves, cabinet feed-throughs or earthing cables to allow you to customize your SymLine™ system
- All accessories are electrically dissipative and approved for use in laboratories with explosion hazards
Specifications
Silver | |
Pipe Mounting | |
Pipe System | |
Mounting Bracket |
32 mm | |
SymLine™ System Customization | |
Metal |