missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Accessories for Horizontal Gel Box, 10-20 cm

For use with horizontal gel box, 10cm.

Brand:  Axygen HGB10-10MC-15

39.71 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 15503107

  • 46.30 € / 1 unit
Estimated Shipment: 15-04-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Combs for 10-20 cm Axygen™ horizontal gel box
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Horizontal Electrophoresis System Accessories
10-well Multi-Channel Compatible Comb, 1.5 mm thickness
1.5 mm
Axygen Horizontal Gel Box, 10 cm
10
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers