missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Accessories for Horizontal Gel Box, 10-20 cm

For use with horizontal gel box, 10cm.

Brand:  Axygen HGB20-25-1

 View more versions of this product

Product Code. 15513157

  • 74.20 € / 1 unit
Estimated Shipment: 10-04-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Combs for 10-20 cm Axygen™ horizontal gel box
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Horizontal Electrophoresis System Accessories
25-well Comb, 1.0 mm thickness
1 mm
Axygen Horizontal Gel Box, 20 cm
25
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers