missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Axygen™ Accessories for Horizontal Gel Box, 10-20 cm
For use with horizontal gel box, 10cm.
Brand: Axygen HGB20-NE
128.10 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
Combs for 10-20 cm Axygen™ horizontal gel boxSpecifications
Horizontal Electrophoresis System Accessories | |
Axygen Horizontal Gel Box, 20 cm | |
Replacement Negative Electrode, 20 cm |