Learn More
Cole-Parmer™ Accessories for Cole-Parmer™ HI-200 Series Hybridization Incubators
Click to view available options
Type:
Tube Rack Holder
Unit Size:
Each
Description
- Expand the capabilities of your Cole Parmer HI-200 Series Hybridization Incubators with our collection of optional accessories
- This collection includes a selection of rocking platforms, hybridization tubes of various sizes, and tube holders with clips
Specifications
Specifications
| For Use With (Equipment) | Hybridization incubator |
| Holds | Up to 3 Large and 3 Small Tubes |
| Material | Plastic |
| Type | Tube Rack Holder |
| Includes | Hybridization Tube Rack Holder |
| Length (Metric) | 80 mm |